• USA Home
  • 8.20535 - N,N-Diphenylhydrazinium chloride

EMAIL THIS PAGE TO A FRIEND
8.20535 EMD Millipore

N,N-Diphenylhydrazinium chloride

N,N-Diphenylhydrazinium chloride for synthesis. CAS 530-47-2, EC Number 208-481-0, chemical formula (C₆H₅)₂NNH₂ * HCl., for synthesis

Synonym: 1,1-Diphenylhydrazine hydrochloride

  • CAS Number 530-47-2

  • Linear Formula C₁₂H₁₃ClN₂

  • Molecular Weight 220.70 g/mol

  •  EC Number 208-481-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8205350010)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?