EMAIL THIS PAGE TO A FRIEND
8.20583 EMD Millipore

Dimethyl fumarate

Dimethyl fumarate for synthesis. CAS 624-49-7, chemical formula CH₃OOCCH=CHCOOCH₃., for synthesis

Synonym: Fumaric acid dimethyl ester

  • CAS Number 624-49-7

  • Linear Formula C₆H₈O₄

  • Molecular Weight 144.13 g/mol

  •  EC Number 210-849-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8205830100)
bp   193 °C (1013 mbar)
vapor pressure   5 hPa (25 °C)
solubility   1.6 g/l
density   1.0283 g/cm3 (20 °C)
mp   102 °C
pH   4.0 - 6.0 (10 g/l, H₂O, 20 °C) suspension
LD 50 oral   LD50 Rat 2240 mg/kg
LD 50 dermal   LD50 Rabbit 1250 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 102 °C
  Melting range (upper value): ≤ 104 °C
  Heavy metals (as Pb): ≤ 0.002 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Hazard Statement(s) 
H312: Harmful in contact with skin.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
irritant, sensitizing
R Phrase 
R 21-36/37/38-43
Harmful in contact with skin.Irritating to eyes, respiratory system and skin.May cause sensitisation by skin contact.
S Phrase 
S 36/37/39
Wear suitable protective clothing, gloves and eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?