• USA Home
  • 8.20595 - 2-Furanmethanethiol

EMAIL THIS PAGE TO A FRIEND
8.20595 EMD Millipore

2-Furanmethanethiol

2-Furanmethanethiol for synthesis. CAS 98-02-2, molar mass 114.16 g/mol., for synthesis

Synonym: Furfuryl mercaptan

  • CAS Number 98-02-2

  • Linear Formula C₅H₆OS

  • Molecular Weight 114.16 g/mol

  •  EC Number 202-628-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8205950100)
  25 ml in Glass bottle (8205950025)
bp   155 °C (1013 hPa)
vapor pressure   4 hPa (20 °C)
density   1.13 g/cm3 (20 °C)
flash point   51 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.126 - 1.128
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P260: Do not breathe vapour.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 2929 , 6.1 (3), II
Declaration (transport by air) IATA-DGR 
UN 2929 , 6.1 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2929 , 6.1 (3), II
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 23-24/25
Do not breathe vapour.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?