• USA Home
  • 8.20618 - O-Benzylhydroxylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.20618 EMD Millipore

O-Benzylhydroxylamine hydrochloride

O-Benzylhydroxylamine hydrochloride for synthesis. CAS 2687-43-6, EC Number 220-249-0, chemical formula C₇H₉NO*HCl., for synthesis

  • CAS Number 2687-43-6

  • Linear Formula C₇H₁₀ClNO

  • Molecular Weight 159.61 g/mol

  •  EC Number 220-249-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8206180010)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Hazard Statement(s) 
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H318: Causes serious eye damage.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
RTECS 
NC3040000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant, sensitizing
R Phrase 
R 38-41-43
Irritating to skin.Risk of serious damage to eyes.May cause sensitisation by skin contact.
S Phrase 
S 24-26-37/39
Avoid contact with skin.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable gloves and eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?