• USA Home
  • 8.20634 - trans, trans-2,4-Hexadienedioic acid

EMAIL THIS PAGE TO A FRIEND
8.20634 EMD Millipore

trans, trans-2,4-Hexadienedioic acid

trans, trans-2,4-Hexadienedioic acid for synthesis. CAS 3588-17-8, EC Number 222-724-8, chemical formula HOOCCH=CHCH=CHCOOH., for synthesis

Synonym: trans, trans-Muconic acid

  • CAS Number 3588-17-8

  • Linear Formula C₆H₆O₄

  • Molecular Weight 142.11 g/mol

  •  EC Number 222-724-8

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   1 g in Glass bottle (8206340001)
solubility   0.2 g/l insoluble
mp   287 - 290 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?