EMAIL THIS PAGE TO A FRIEND
8.20642 EMD Millipore

cis-3-Hexen-1-ol

cis-3-Hexen-1-ol for synthesis. CAS 928-96-1, chemical formula CH₃CH₂CH=CHCH₂CH₂OH., for synthesis

  • CAS Number 928-96-1

  • Linear Formula C₆H₁₂O

  • Molecular Weight 100.16 g/mol

  •  EC Number 213-192-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8206420025)
bp   156 - 157 °C (1013 hPa)
density   0.85 g/cm3 (20 °C)
flash point   44 °C (External MSDS)
LD 50 oral   LD50 Rat 4700 mg/kg
LD 50 dermal   LD50 Rat > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.848 - 0.850
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 29 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?