EMAIL THIS PAGE TO A FRIEND
8.20645 EMD Millipore

3-Hexyne

3-Hexyne for synthesis. CAS 928-49-4, molar mass 82.15 g/mol., for synthesis

  • CAS Number 928-49-4

  • Linear Formula C₆H₁₀

  • Molecular Weight 82.15 g/mol

  •  EC Number 213-173-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8206450010)
bp   81 - 82 °C
vapor pressure   248 hPa (37.7 °C)
solubility   0.559 g/l
density   0.72 g/cm3 (20 °C)
flash point   -14 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 98.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.723 - 0.724
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2901 29 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P223: Keep away from any possible contact with water, because of violent reaction and possible flash fire.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable, irritant
R Phrase 
R 11-36/37/38
Highly flammable.Irritating to eyes, respiratory system and skin.
S Phrase 
S 16-26-33
Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Take precautionary measures against static discharges.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?