• USA Home
  • 8.20737 - o-Iodohippuric acid sodium salt dihydrate

EMAIL THIS PAGE TO A FRIEND
8.20737 EMD Millipore

o-Iodohippuric acid sodium salt dihydrate

o-Iodohippuric acid sodium salt dihydrate for synthesis. CAS 5990-94-3, EC Number 205-097-5, chemical formula 2-(I)C₆H₄CONHCH₂COONa * 2H₂O., for synthesis

Synonym: (2-Iodobenzoyl)glycine sodium salt

  • CAS Number 5990-94-3

  • Linear Formula C₉H₇INNaO₃ * 2H₂O

  • Molecular Weight 327.06 g/mol

  •  EC Number 205-097-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8207370010)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 98
  Identity (IR): passes test
  o- Iodobenzoic acid (HPLC, contra external standard): ≤ 0.1 % (w/w)

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?