EMAIL THIS PAGE TO A FRIEND
8.20739 EMD Millipore

1-Iodooctane

1-Iodooctane for synthesis. CAS 629-27-6, molar mass 240.13 g/mol., for synthesis

Synonym: Octyl iodide

  • CAS Number 629-27-6

  • Linear Formula C₈H₁₇I

  • Molecular Weight 240.13 g/mol

  •  EC Number 211-084-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8207390050)
bp   225 - 226 °C (1013 hPa)
density   1.32 g/cm3 (20 °C)
flash point   87 °C
mp   -46 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.320 - 1.335
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 80
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
RG9700100
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?