EMAIL THIS PAGE TO A FRIEND
8.20741 EMD Millipore

2-Iodophenol

2-Iodophenol for synthesis. CAS 533-58-4, chemical formula 2-(I)C₆H₄OH., for synthesis

Synonym: 2-Iodo-1-hydroxybenzene

  • CAS Number 533-58-4

  • Linear Formula C₆H₅IO

  • Molecular Weight 220.01 g/mol

  •  EC Number 208-569-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8207410001)
bp   186 - 187 °C (213 hPa)
mp   39 - 42 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 39 °C
  Melting range (upper value): ≤ 42 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2908 19 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?