EMAIL THIS PAGE TO A FRIEND
8.20845 EMD Millipore

2-Naphthaldehyde

2-Naphthaldehyde for synthesis. CAS 66-99-9, molar mass 156.18 g/mol., for synthesis

Synonym: 2-Naphthalenecarbaldehyde

  • CAS Number 66-99-9

  • Linear Formula C₁₁H₈O

  • Molecular Weight 156.18 g/mol

  •  EC Number 200-640-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8208450010)
flash point   >100 °C
mp   57 - 60 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 57 °C
  Melting range (upper value): ≤ 60 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
RTECS 
QJ0190010
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?