EMAIL THIS PAGE TO A FRIEND
8.20858 EMD Millipore

2-Naphthalenethiol

2-Naphthalenethiol for synthesis. CAS 91-60-1, molar mass 160.24 g/mol., for synthesis

Synonym: 2-Thionaphthol, 2-Naphthyl mercaptan

  • CAS Number 91-60-1

  • Linear Formula C₁₀H₈S

  • Molecular Weight 160.24 g/mol

  •  EC Number 202-082-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8208580010)
bp   286 °C (1013 hPa)
density   1.55 g/cm3 (20 °C)
mp   79 - 82 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 78 °C
  Melting range (upper value): ≤ 81 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
UN 3335 , 9, III
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?