• USA Home
  • 8.20878 - Phenylmercury nitrate (basic)

EMAIL THIS PAGE TO A FRIEND
8.20878 EMD Millipore

Phenylmercury nitrate (basic)

for synthesis

Synonym: Nitratophenylmercury

  • CAS Number 8003-05-2

  • Linear Formula C₁₂H₁₁Hg₂NO₄

  • Molecular Weight 634.4 g/mol

Purchase

Properties

packaging   50 g in Glass bottle (8208780050)
mp   187 - 190 °C (decomposition)
pH   6.6 (10 g/l, H₂O, 20 °C)
bulk density   800 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex Hg): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2852 10 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H314: Causes severe skin burns and eye damage.
H372: Causes damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
OW8575000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
20
Mercury residues and salts. Elemental mercury must be taken up with Chemizorb® Hg (Cat. No 112576): container G.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 1895 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 1895 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 1895 , 6.1, II, Marine Pollutant: P, Segregation Group: 7 (Heavy metals and their salts (including their organometallic compounds))11 (Mercury and mercury compounds)
Categories of danger 
toxic, corrosive, dangerous for the environment
R Phrase 
R 25-34-48/24/25-50/53
Toxic if swallowed.Causes burns.Toxic: danger of serious damage to health by prolonged exposure in contact with skin and if swallowed.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 23-24/25-37-45-60-61
Do not breathe vapour.Avoid contact with skin and eyes.Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).This material and its container must be disposed of as hazardous waste.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?