EMAIL THIS PAGE TO A FRIEND
8.20882 EMD Millipore

3-Nitroanisole

3-Nitroanisole for synthesis. CAS No. 555-03-3, EC Number 209-079-8., for synthesis

Synonym: 3-Methoxynitrobenzene

  • CAS Number 555-03-3

  • Linear Formula C₇H₇NO₃

  • Molecular Weight 153.14 g/mol

  •  EC Number 209-079-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8208820050)
bp   256.3 °C (1013 hPa)
solubility   500 mg/l
flash point   127.9 °C (External MSDS)
mp   34 - 37 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 34 °C
  Melting range (upper value): ≤ 37 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 3458 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3458 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3458 , 6.1, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?