• USA Home
  • 8.20888 - 2-Nitrobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.20888 EMD Millipore

2-Nitrobenzyl alcohol

2-Nitrobenzyl alcohol for synthesis. CAS No. 612-25-9, EC Number 210-302-6., for synthesis

  • CAS Number 612-25-9

  • Linear Formula C₇H₇NO₃

  • Molecular Weight 153.14 g/mol

  •  EC Number 210-302-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8208880025)
bp   270 °C
mp   70 - 73 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 70 °C
  Melting range (upper value): ≤ 73 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?