• USA Home
  • 8.20889 - 4-Nitrobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.20889 EMD Millipore

4-Nitrobenzyl alcohol

4-Nitrobenzyl alcohol for synthesis. CAS 619-73-8, chemical formula 4-(NO₂)C₆H₄CH₂OH., for synthesis

  • CAS Number 619-73-8

  • Linear Formula C₇H₇NO₃

  • Molecular Weight 153.14 g/mol

  •  EC Number 210-611-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8208890025)
bp   185 °C (16 hPa)
solubility   2 g/l
flash point   180 °C
mp   90 - 93 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 92 °C
  Melting range (upper value): ≤ 95 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?