• USA Home
  • 8.20892 - 4-Nitropyrocatechol

EMAIL THIS PAGE TO A FRIEND
8.20892 EMD Millipore

4-Nitropyrocatechol

for synthesis

  • CAS Number 3316-09-4

  • Linear Formula C₆H₅NO₄

  • Molecular Weight 155.11 g/mol

  •  EC Number 222-009-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8208920005)
mp   173 - 177 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 97.0 %
  Melting range (lower value): ≥ 173 °C
  Melting range (upper value): ≤ 177 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2908 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?