• USA Home
  • 8.20897 - (4-Nitrophenyl)acetonitrile

EMAIL THIS PAGE TO A FRIEND
8.20897 EMD Millipore

(4-Nitrophenyl)acetonitrile

(4-Nitrophenyl)acetonitrile for synthesis. CAS No. 555-21-5, EC Number 209-085-0., for synthesis

Synonym: 4-Nitrobenzyl cyanide

  • CAS Number 555-21-5

  • Linear Formula C₈H₆N₂O₂

  • Molecular Weight 162.14 g/mol

  •  EC Number 209-085-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8208970050)
bp   195 - 197 °C (16 hPa)
mp   112 - 115 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 112 °C
  Melting range (upper value): ≤ 115 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?