EMAIL THIS PAGE TO A FRIEND
8.20974 EMD Millipore

m-Phenetidine

m-Phenetidine for synthesis. CAS 621-33-0, EC Number 210-680-2, chemical formula 3-(C₂H₅O)C₆H₄NH₂., for synthesis

Synonym: 3-Ethoxyaniline, 3'-Aminophenetol

  • CAS Number 621-33-0

  • Linear Formula C₈H₁₁NO

  • Molecular Weight 137.18 g/mol

  •  EC Number 210-680-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8209740050)
bp   248 °C (1013 hPa)
vapor pressure   30 hPa (50 °C)
density   1.07 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.065 - 1.067
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 29 00
Hazard Statement(s) 
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled.
H373: May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
SI5400000
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2311 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2311 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2311 , 6.1, III
Categories of danger 
toxic
R Phrase 
R 23/24/25-33
Toxic by inhalation, in contact with skin and if swallowed.Danger of cumulative effects.
S Phrase 
S 36/37-45
Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?