• USA Home
  • 8.21002 - Diethyl phenylmalonate

EMAIL THIS PAGE TO A FRIEND
8.21002 EMD Millipore

Diethyl phenylmalonate

Diethyl phenylmalonate for synthesis. CAS 83-13-6, chemical formula C₆H₅CH(COOC₂H₅)₂., for synthesis

Synonym: Phenylmalonic acid diethyl ester

  • CAS Number 83-13-6

  • Linear Formula C₁₃H₁₆O₄

  • Molecular Weight 236.26 g/mol

  •  EC Number 201-456-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8210020005)
bp   158 - 159 °C (13 hPa)
density   1.10 g/cm3 (20 °C)
flash point   113 °C
mp   16 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.095 - 1.097
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 39 95
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?