• USA Home
  • 8.21013 - Ethyl phenylacetylenecarboxylate

EMAIL THIS PAGE TO A FRIEND
8.21013 EMD Millipore

Ethyl phenylacetylenecarboxylate

Ethyl phenylacetylenecarboxylate for synthesis. CAS 2216-94-6, molar mass 174.20 g/mol., for synthesis

Synonym: Phenylacetylenecarboxylic acid ethyl ester, Ethyl phenylpropiolate

  • CAS Number 2216-94-6

  • Linear Formula C₁₁H₁₀O₂

  • Molecular Weight 174.20 g/mol

  •  EC Number 218-703-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8210130005)
bp   128 °C (13 hPa)
density   1.05 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.056 - 1.058
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
RTECS 
UE0110000
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?