EMAIL THIS PAGE TO A FRIEND
8.21018 EMD Millipore

Diphenyl sulfoxide

Diphenyl sulfoxide for synthesis. CAS 945-51-7, chemical formula (C₆H₅)₂SO., for synthesis

Synonym: Phenyl sulfoxide

  • CAS Number 945-51-7

  • Linear Formula C₁₂H₁₀OS

  • Molecular Weight 202.27 g/mol

  •  EC Number 213-415-9

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfoxides,
packaging   25 g in Plastic bottle (8210180025)
bp   206 - 208 °C (7 hPa)
mp   67 - 71 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 67 °C
  Melting range (upper value): ≤ 71 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?