• USA Home
  • 8.21217 - (2R,3R)-(+)-Diethyl tartrate

EMAIL THIS PAGE TO A FRIEND
8.21217 EMD Millipore

(2R,3R)-(+)-Diethyl tartrate

(2R,3R)-(+)-Diethyl tartrate for synthesis. CAS 87-91-2, chemical formula C₂H₅OOCCH(OH)CH(OH)COOC₂H₅., for synthesis

Synonym: L-(+)-Tartaric acid diethyl ester, DET, (+)-Diethyl-L-tartrate

  • CAS Number 87-91-2

  • Linear Formula C₈H₁₄O₆

  • Molecular Weight 206.19 g/mol

  •  EC Number 201-783-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8212170050)
bp   153 - 156 °C (16 hPa)
density   1.21 g/cm3 (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.204 - 1.207
  Spec. rotation [α²0/D (undiluted): +7.5 - +9.5 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 13 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?