• USA Home
  • 8.21223 - 2,4-Dimethylaniline

EMAIL THIS PAGE TO A FRIEND
8.21223 EMD Millipore

2,4-Dimethylaniline

2,4-Dimethylaniline for synthesis. CAS 95-68-1, chemical formula 2,4-(CH₃)₂C₆H₃NH₂., for synthesis

Synonym: asym.-m-Xylidine

  • CAS Number 95-68-1

  • Linear Formula C₈H₁₁N

  • Molecular Weight 121.18 g/mol

  •  EC Number 202-440-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8212230005)
bp   214 °C (1013 hPa)
expl. lim.     1.1 - 7.0 %(V)
vapor pressure   0.11 hPa (20 °C)
solubility   5 g/l
density   0.98 g/cm3 (20 °C)
flash point   98 °C
autoignition temp.   520 °C
mp   -16 °C
pH   7 (22 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.975 - 0.977
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H331: Toxic if inhaled.
H311: Toxic in contact with skin.
H301: Toxic if swallowed.
H411: Toxic to aquatic life with long lasting effects.
H373: May cause damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 1711 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 1711 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 1711 , 6.1, II
Categories of danger 
toxic, dangerous for the environment
R Phrase 
R 23/24/25-33-51/53
Toxic by inhalation, in contact with skin and if swallowed.Danger of cumulative effects.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 28-36/37-45-61
After contact with skin, wash immediately with plenty of soap and water.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?