• USA Home
  • 8.21226 - 2-Methoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.21226 EMD Millipore

2-Methoxybenzyl alcohol

2-Methoxybenzyl alcohol for synthesis. CAS 612-16-8, chemical formula 2-(CH₃O)C₆H₄CH₂OH., for synthesis

Synonym: o-Anise alcohol

  • CAS Number 612-16-8

  • Linear Formula C₈H₁₀O₂

  • Molecular Weight 138.17 g/mol

  •  EC Number 210-296-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8212260025)
bp   248 - 250 °C
density   1.12 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.122 - 1.123
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 49 80
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?