• USA Home
  • 8.21227 - 3-Methoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.21227 EMD Millipore

3-Methoxybenzyl alcohol

3-Methoxybenzyl alcohol for synthesis. CAS No. 6971-51-3, EC Number 230-200-5., for synthesis

Synonym: m-Anise alcohol

  • CAS Number 6971-51-3

  • Linear Formula C₈H₁₀O₂

  • Molecular Weight 138.17 g/mol

  •  EC Number 230-200-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8212270025)
bp   105 - 110 °C (4 hPa)
density   1.11 g/cm3 (20 °C)
flash point   >100 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.114 - 1.115
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 49 80
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?