EMAIL THIS PAGE TO A FRIEND
8.21232 EMD Millipore

3-Methoxyphenol

3-Methoxyphenol for synthesis. CAS No. 150-19-6, EC Number 205-754-6., for synthesis

Synonym: 1-Hydroxy-3-methoxybenzene, 3-Hydroxyanisole, 97%, Resorcinol monomethyl ether

  • CAS Number 150-19-6

  • Linear Formula C₇H₈O₂

  • Molecular Weight 124.14 g/mol

  •  EC Number 205-754-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8212320050)
bp   113 - 115 °C (7 hPa)
density   1.15 g/cm3 (20 °C)
flash point   124 °C
mp   -18 - -16 °C
LD 50 oral   LD50 Rat 597 mg/kg
LD 50 dermal   LD50 Rat 682 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.145 - 1.147
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 50 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H311: Toxic in contact with skin.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
Categories of danger 
harmful, irritant
R Phrase 
R 21/22-36/38
Harmful in contact with skin and if swallowed.Irritating to eyes and skin.
S Phrase 
S 26-36/37
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?