• USA Home
  • 8.21602 - Monomethyl terephthalate

EMAIL THIS PAGE TO A FRIEND
8.21602 EMD Millipore

Monomethyl terephthalate

Monomethyl terephthalate for synthesis. CAS 1679-64-7, EC Number 216-849-7, chemical formula 4-(HOOC)C₆H₄COOCH₃., for synthesis

Synonym: Terephthalic acid monomethyl ester

  • CAS Number 1679-64-7

  • Linear Formula C₉H₈O₄

  • Molecular Weight 180.16 g/mol

  •  EC Number 216-849-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8216020010)
solubility   0.13 g/l
mp   220 - 223 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 95.0 % (a/a)
  Melting range (lower value): ≥ 215 °C
  Melting range (upper value): ≤ 220 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 39 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?