EMAIL THIS PAGE TO A FRIEND
8.21668 EMD Millipore

Allylbenzene

Allylbenzene for synthesis. CAS No. 300-57-2, EC Number 206-095-7., for synthesis

Synonym: 3-Phenyl-1-propene

  • CAS Number 300-57-2

  • Linear Formula C₉H₁₀

  • Molecular Weight 118.17 g/mol

  •  EC Number 206-095-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8216680010)
bp   156 - 157 °C (1013 hPa)
density   0.89 g/cm3 (20 °C)
flash point   40 °C
mp   <-40 °C
LD 50 oral   LD50 Rat 5540 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.887 - 0.897
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 90 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P260: Do not breathe vapour.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 23-24/25
Do not breathe vapour.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?