• USA Home
  • 8.21675 - 2-Amino-4-methylphenol

EMAIL THIS PAGE TO A FRIEND
8.21675 EMD Millipore

2-Amino-4-methylphenol

2-Amino-4-methylphenol for synthesis. CAS 95-84-1, chemical formula 2-(NH₂)-4-(CH₃)C₆H₃OH., for synthesis

Synonym: 2-Amino-p-cresol, 3-Amino-4-hydroxytoluene, 2-Hydroxy-5-methylaniline

  • CAS Number 95-84-1

  • Linear Formula C₇H₉NO

  • Molecular Weight 123.15 g/mol

  •  EC Number 202-457-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8216750100)
mp   133 - 136 °C
bulk density   500 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 133 °C
  Melting range (upper value): ≤ 136 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 29 00
RTECS 
SJ6078000
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2512 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2512 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2512 , 6.1, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?