• USA Home
  • 8.21699 - 3-Bromobenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.21699 EMD Millipore

3-Bromobenzonitrile

3-Bromobenzonitrile for synthesis. CAS No. 6952-59-6, EC Number 230-127-9., for synthesis

  • CAS Number 6952-59-6

  • Linear Formula C₇H₄BrN

  • Molecular Weight 182.01 g/mol

  •  EC Number 230-127-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8216990010)
bp   225 °C (1013 hPa)
solubility   0.2 g/l
density   1.562 g/cm3 (20 °C)
flash point   113 °C
mp   37 - 40 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 37 °C
  Melting range (upper value): ≤ 40 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
H319: Causes serious eye irritation.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, III
Categories of danger 
harmful, irritant, dangerous for the environment
R Phrase 
R 21/22-36-52/53
Harmful in contact with skin and if swallowed.Irritating to eyes.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 36/37-61
Wear suitable protective clothing and gloves.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?