EMAIL THIS PAGE TO A FRIEND
8.21707 EMD Millipore

Bromohexane

Bromohexane (mixtures of isomeres) for synthesis. CAS No. 3377-86-4, EC Number 222-173-3., (mixtures of isomeres) for synthesis

Synonym: Hexyl bromide

  • CAS Number 3377-86-4

  • Linear Formula C₆H₁₃Br

  • Molecular Weight 165.08 g/mol

  •  EC Number 222-173-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8217070025)
bp   142 - 144 °C (1013 hPa)
vapor pressure   9.7 hPa (25 °C)
density   1.166 g/cm3 (20 °C)
flash point   34 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay isomers (GC, area%): ≥ 99.0 % (a/a)
  3-isomer (GC, area%): 22.0 - 28.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.162 - 1.167
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 19
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?