• USA Home
  • 8.21709 - 3-(Bromomethyl)heptane

EMAIL THIS PAGE TO A FRIEND
8.21709 EMD Millipore

3-(Bromomethyl)heptane

3-(Bromomethyl)heptane for synthesis. CAS No. 18908-66-2, EC Number 242-659-9., for synthesis

Synonym: 2-Ethylhexyl bromide, Isooctyl bromide, 1-Bromo-2-ethylhexane

  • CAS Number 18908-66-2

  • Linear Formula C₈H₁₇Br

  • Molecular Weight 193.12 g/mol

  •  EC Number 242-659-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8217090100)
bp   75 - 77 °C (21 hPa)
density   1.12 g/cm3 (20 °C)
flash point   70 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.118 - 1.125
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 19
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?