EMAIL THIS PAGE TO A FRIEND
8.21714 EMD Millipore

4-Bromophenol

4-Bromophenol for synthesis. CAS 106-41-2, molar mass 173 g/mol., for synthesis

Synonym: 1-Brom-1-hydroxybenzene

  • CAS Number 106-41-2

  • Linear Formula C₆H₅BrO

  • Molecular Weight 173 g/mol

  •  EC Number 203-394-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8217140100)
  500 g in Glass bottle (8217140500)
bp   238 °C (1013 hPa)
solubility   14 g/l
flash point   >110 °C
mp   64 - 66 °C
shipped in   ambient
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 63 °C
  Melting range (upper value): ≤ 66 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2908 19 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
SJ7960000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-38
Harmful if swallowed.Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?