EMAIL THIS PAGE TO A FRIEND
8.21715 EMD Millipore

2-Bromotoluene

2-Bromotoluene for synthesis. CAS 95-46-5, molar mass 171.03 g/mol., for synthesis

  • CAS Number 95-46-5

  • Linear Formula C₇H₇Br

  • Molecular Weight 171.03 g/mol

  •  EC Number 202-421-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8217150025)
bp   179 - 181 °C (1013 hPa)
vapor pressure   1 hPa (25 °C)
density   1.42 g/cm3 (20 °C)
flash point   79 °C
autoignition temp.   >100 °C
mp   -27 °C
LD 50 oral   LD50 Rat 1540 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.418 - 1.428
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H302: Harmful if swallowed.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
RTECS 
XS7965500
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
Categories of danger 
harmful, dangerous for the environment
R Phrase 
R 22-51/53
Harmful if swallowed.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24/25-61
Avoid contact with skin and eyes.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?