• USA Home
  • 8.21798 - 1-Fluoro-2-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
8.21798 EMD Millipore

1-Fluoro-2-nitrobenzene

1-Fluoro-2-nitrobenzene for synthesis. CAS 1493-27-2, chemical formula 1-(F)C₆H₄-2-(NO₂)., for synthesis

Synonym: o-Fluornitrobenzene

  • CAS Number 1493-27-2

  • Linear Formula C₆H₄FNO₂

  • Molecular Weight 141.1 g/mol

  •  EC Number 216-088-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8217980025)
bp   215 °C (1013 hPa)
density   1.33 g/cm3 (20 °C)
flash point   94 °C
mp   -8 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.333 - 1.335
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?