EMAIL THIS PAGE TO A FRIEND
8.21802 EMD Millipore

4-Fluorotoluene

4-Fluorotoluene for synthesis. CAS 352-32-9, chemical formula 4-(F)C₆H₄CH₃., for synthesis

  • CAS Number 352-32-9

  • Linear Formula C₇H₇F

  • Molecular Weight 110.13 g/mol

  •  EC Number 206-520-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8218020100)
bp   116 °C
vapor pressure   23 hPa (20 °C)
solubility   200 mg/l
density   1.00 g/cm3 (20 °C)
flash point   10 °C
mp   -57 °C
LD 50 oral   LD50 Rat 7000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.996 - 0.998
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
P240: Ground/bond container and receiving equipment.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 2388 , 3, II
Declaration (transport by air) IATA-DGR 
UN 2388 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 2388 , 3, II, Segregation Group: 10 (Liquid halogenated hydrocarbons)
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16-24/25
Keep away from sources of ignition - No smoking.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?