• USA Home
  • 8.21825 - 1-Iodo-3-methylbutane

EMAIL THIS PAGE TO A FRIEND
8.21825 EMD Millipore

1-Iodo-3-methylbutane

1-Iodo-3-methylbutane (stabilised with silver) for synthesis. CAS 541-28-6, chemical formula (CH₃)₂CHCH₂CH₂I., (stabilised with silver) for synthesis

Synonym: Isoamyl iodide, Isopentyl iodide

  • CAS Number 541-28-6

  • Linear Formula C₅H₁₁I

  • Molecular Weight 198.04 g/mol

  •  EC Number 208-773-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8218250025)
bp   146 - 148 °C (1013 hPa)
density   1.50 g/cm3 (20 °C)
flash point   40 °C
LD 50 oral   LD50 Rat 1424 mg/kg
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302: Harmful if swallowed.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable, harmful, dangerous for the environment
R Phrase 
R 10-22
Flammable.Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?