• USA Home
  • 8.21855 - Ethyl-4-nitrocinnamate

EMAIL THIS PAGE TO A FRIEND
8.21855 EMD Millipore

Ethyl-4-nitrocinnamate

Ethyl-4-nitrocinnamate for synthesis. CAS 953-26-4, EC Number 213-463-0, chemical formula 4-(NO₂)C₆H₄CH=CHCOOC₂H₅., for synthesis

Synonym: 4-Nitrocinnamic acid ethyl ester

  • CAS Number 953-26-4

  • Linear Formula C₁₁H₁₁NO₄

  • Molecular Weight 221.21 g/mol

  •  EC Number 213-463-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8218550010)
density   0.87 g/cm3 (20 °C)
mp   138 - 140 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 135 °C
  Melting range (upper value): ≤ 138 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?