EMAIL THIS PAGE TO A FRIEND
8.21856 EMD Millipore

1-Nonadecanol

1-Nonadecanol for synthesis. CAS 1454-84-8, chemical formula CH₃(CH₂)₁₇CH₂OH., for synthesis

Synonym: N-Nonadecyl alcohol

  • CAS Number 1454-84-8

  • Linear Formula C₁₉H₄₀O

  • Molecular Weight 284.52 g/mol

  •  EC Number 215-930-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8218560001)
bp   156 °C (13.3 hPa)
flash point   >110 °C
mp   60 - 61 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.5 % (a/a)
  Melting range (lower value): ≥ 58 °C
  Melting range (upper value): ≤ 61 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?