EMAIL THIS PAGE TO A FRIEND
8.21859 EMD Millipore

3-Octanol

3-Octanol for synthesis. CAS No. 589-98-0, EC Number 209-667-4., for synthesis

Synonym: Ethyl pentyl carbinol

  • CAS Number 589-98-0

  • Linear Formula C₈H₁₈O

  • Molecular Weight 130.22 g/mol

  •  EC Number 209-667-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8218590050)
bp   175 °C (1013 hPa)
density   0.82 g/cm3 (20 °C)
flash point   68 °C
mp   -45 °C
LD 50 oral   LD50 Rat > 5000 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.822 - 0.824
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 16 85
Hazard Statement(s) 
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36
Irritating to eyes.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?