EMAIL THIS PAGE TO A FRIEND
8.21860 EMD Millipore

3-Octanone

3-Octanone for synthesis. CAS No. 106-68-3, EC Number 203-423-0., for synthesis

Synonym: Ethyl pentyl ketone, Ethyl n-amyl ketone, EAK, n-Amyl ethyl ketone

  • CAS Number 106-68-3

  • Linear Formula C₈H₁₆O

  • Molecular Weight 128.21 g/mol

  •  EC Number 203-423-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8218600100)
bp   169 °C (1013 hPa)
vapor pressure   2 hPa (20 °C)
solubility   2.60 g/l
density   0.82 g/cm3 (20 °C)
flash point   48 °C
autoignition temp.   330 °C
mp   -23 °C
LD 50 oral   LD50 Rat > 3500 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.820 - 0.821
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 19 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 2271 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2271 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2271 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?