• USA Home
  • 8.21876 - 3-Methylpyridine 1-oxide

EMAIL THIS PAGE TO A FRIEND
8.21876 EMD Millipore

3-Methylpyridine 1-oxide

3-Methylpyridine 1-oxide for synthesis. CAS 1003-73-2, molar mass 109.13 g/mol., for synthesis

Synonym: 3-Picoline N-oxide

  • CAS Number 1003-73-2

  • Linear Formula C₆H₇NO

  • Molecular Weight 109.13 g/mol

  •  EC Number 213-714-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8218760100)
bp   150 °C (20 hPa)
vapor pressure   0.01 hPa (20 °C)
solubility   >2000 g/l
density   1.13 g/cm3 (40 °C)
flash point   155 °C
mp   37 - 38 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 36 °C
  Melting range (upper value): ≤ 39 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
UT5799000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?