• USA Home
  • 8.21890 - 1,2-Bis(dibromomethyl)-benzene

EMAIL THIS PAGE TO A FRIEND
8.21890 EMD Millipore

1,2-Bis(dibromomethyl)-benzene

1,2-Bis(dibromomethyl)-benzene for synthesis. CAS 13209-15-9, chemical formula C₆H₄-1,2-(CHBr₂)₂., for synthesis

Synonym: α,α,α',α'-Tetrabromo-o-xylene

  • CAS Number 13209-15-9

  • Linear Formula C₈H₆Br₄

  • Molecular Weight 421.75 g/mol

  •  EC Number 236-176-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   117 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3143 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3143 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3143 , 6.1, II
Categories of danger 
irritant, dangerous for the environment
R Phrase 
R 36/38-50/53
Irritating to eyes and skin.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?