• USA Home
  • 8.21964 - Bis(4-chlorophenyl) sulfone

EMAIL THIS PAGE TO A FRIEND
8.21964 EMD Millipore

Bis(4-chlorophenyl) sulfone

Bis(4-chlorophenyl) sulfone for synthesis. CAS No. 80-07-9, EC Number 201-247-9., for synthesis

Synonym: 4,4'-Dichlorodiphenyl sulfone

  • CAS Number 80-07-9

  • Linear Formula C₁₂H₈Cl₂O₂S

  • Molecular Weight 287.16 g/mol

  •  EC Number 201-247-9

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfones,
packaging   250 g in Plastic bottle (8219640250)
bp   250 °C (13 hPa)
mp   144 - 148 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex Cl): ≥ 98.0 %
  Melting range (lower value): ≥ 144 °C
  Melting range (upper value): ≤ 148 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?