• USA Home
  • 8.21969 - 1-Bromo-4-chlorobenzene

EMAIL THIS PAGE TO A FRIEND
8.21969 EMD Millipore

1-Bromo-4-chlorobenzene

1-Bromo-4-chlorobenzene for synthesis. CAS 106-39-8, chemical formula 1-(Br)-4-(Cl)C₆H₄., for synthesis

  • CAS Number 106-39-8

  • Linear Formula C₆H₄BrCl

  • Molecular Weight 191.45 g/mol

  •  EC Number 203-392-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8219690005)
bp   196 °C (1013 hPa)
solubility   0.045 g/l
mp   64 - 67 °C
bulk density   900 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 64 °C
  Melting range (upper value): ≤ 67 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
RTECS 
CY9006000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?