EMAIL THIS PAGE TO A FRIEND
8.21981 EMD Millipore

Isopropyl butyrate

Isopropyl butyrate for synthesis. CAS 638-11-9, EC Number 211-320-7, chemical formula CH₃CH₂CH₂COOCH(CH₃)₂., for synthesis

Synonym: Butyric acid isopropyl ester

  • CAS Number 638-11-9

  • Linear Formula C₇H₁₄O₂

  • Molecular Weight 130.19 g/mol

  •  EC Number 211-320-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   130 °C (1013 hPa)
vapor pressure   65 hPa (50 °C)
density   0.86 g/cm3 (20 °C)
flash point   24 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.860 - 0.865
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 60 19
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 2405 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2405 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2405 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?