• USA Home
  • 8.22105 - 2'-Nitroacetophenone

EMAIL THIS PAGE TO A FRIEND
8.22105 EMD Millipore

2'-Nitroacetophenone

2'-Nitroacetophenone for synthesis. CAS 577-59-3, chemical formula 2-(NO₂)C₆H₄COCH₃., for synthesis

Synonym: 2-Nitro-1-acetylbenzene

  • CAS Number 577-59-3

  • Linear Formula C₈H₇NO₃

  • Molecular Weight 165.15 g/mol

  •  EC Number 209-414-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8221050010)
bp   159 °C (22 hPa)
density   1.24 g/cm3 (20 °C)
flash point   112 °C
mp   27 - 28 °C
LD 50 oral   LD50 Rat 1600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.239 - 1.245
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?