• USA Home
  • 8.22106 - 3-Nitrobenzoic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
8.22106 EMD Millipore

3-Nitrobenzoic acid sodium salt

3-Nitrobenzoic acid sodium salt for synthesis. CAS No. 827-95-2, EC Number 212-578-3., for synthesis

  • CAS Number 827-95-2

  • Linear Formula C₇H₄NNaO₄

  • Molecular Weight 189.1 g/mol

  •  EC Number 212-578-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8221060100)
solubility   10.9 g/l
bulk density   620 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex Na, calc. on anhydrous substance): ≥ 98.0 %
  Water (K. F.): ≤ 1.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?