EMAIL THIS PAGE TO A FRIEND
8.22111 EMD Millipore

Methyl nonanoate

Methyl nonanoate for synthesis. CAS 1731-84-6, EC Number 217-052-7, chemical formula CH₃(CH₂)₇COOCH₃., for synthesis

Synonym: Methyl pelargonate, Pelargonic acid methyl ester, Nonanoic acid methyl ester

  • CAS Number 1731-84-6

  • Linear Formula C₁₀H₂₀O₂

  • Molecular Weight 172.27 g/mol

  •  EC Number 217-052-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8221110250)
density   0.88 g/cm3 (20 °C)
flash point   87 °C
mp   -30 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.874 - 0.876
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?